missing translation for 'onlineSavingsMsg'
Learn More

Amitriptyline hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019569
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019569 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019569 Supplier Merck Emd Millipore Supplier No. PHR13841G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Amitriptyline hydrochloride is a drug, and a tricyclic antidepressant with anticholinergic activity. It is widely used in combination with chlordiazepoxide 7-chloro-2-(methyl-amino)-5-phenyl-3H-1,4-benzodiazepine-4-oxide for the treatment of patients with moderate to severe depression associated with moderate to severe anxiety. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 549-18-8
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C20H23 N · HCl
MDL Number MFCD00012537
Synonym Amitriptyline hydrochloride
Type Certified Reference Material
Quantity 1 g
Formula Weight 313.86
Grade Certified Reference Material