missing translation for 'onlineSavingsMsg'
Learn More

Amitraz 97.0+%, TCI America™

Catalog No. p-7117989
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2608250MG 250 mg
A26081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2608250MG Supplier TCI America Supplier No. A2608250MG
Only null left

Chemical Identifiers

CAS 33089-61-1
Molecular Formula C19H23N3
Molecular Weight (g/mol) 293.414
MDL Number MFCD00069396
InChI Key QXAITBQSYVNQDR-UHFFFAOYSA-N
Synonym amitraz, mitac, taktic, triazid, azaform, mitaban, acarac, baam, azadieno, ectodex
PubChem CID 36324
ChEBI CHEBI:2665
IUPAC Name N'-(2,4-dimethylphenyl)-N-[(2,4-dimethylphenyl)iminomethyl]-N-methylmethanimidamide
SMILES CC1=CC(=C(C=C1)N=CN(C)C=NC2=C(C=C(C=C2)C)C)C

Specifications

Melting Point 86°C
Color White-Yellow
Quantity 250 mg
Physical Form Crystal-Powder at 20°C
UN Number 3077
Formula Weight 293.41
Percent Purity ≥97.0% (GC,T)
Chemical Name or Material Amitraz
RTECSNumber ZF0480000
TSCA No
Recommended Storage Freezer