missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amitraz 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 86°C |
| Color | White-Yellow |
| Quantity | 250 mg |
| Physical Form | Crystal-Powder at 20°C |
| UN Number | 3077 |
| Formula Weight | 293.41 |
| Percent Purity | ≥97.0% (GC,T) |
| Chemical Name or Material | Amitraz |
Safety and Handling
| RTECSNumber | ZF0480000 |
| TSCA | No |
| Recommended Storage | Freezer |