missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amiodarone hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Ampule of 1 mL, Certified Reference Material, Cerilliant™
Description
- Amiodarone, marketed as Pacerone™, Cordarone™, Aratac, and Atlansil, is an antiarrhythmic agent monitored by clinical labs using HPLC or LCMS to ensure patients remain within the drugs therapeutic range.
Specifications
Specifications
| Chemical Name or Material | Amiodarone HCl |
| Packaging | Ampule |
| CAS | 19774-82-4 |
| Molecular Formula | C25H30ClI2NO3 |
| Concentration | 1.0 mg/mL |
| For Use With (Equipment) | Liquid Chromatography (LC); Gas Chromatography (GC) |
| Solution Type | Standard Solution |
| Type | Certified Reference Material |
| Industry Type | Clinical Testing |
| Molecular Weight (g/mol) | 681.77 |
| Show More |