missing translation for 'onlineSavingsMsg'
Learn More

Amiodarone hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111017845
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017845 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017845 Supplier Merck Emd Millipore Supplier No. PHR11641G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Amiodarone hydrochloride is an antiarrhythmic drug used for the treatment of ventricular and supraventricular tachycardia. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 19774-82-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C25H29I2 NO3 · HCl
MDL Number MFCD00069204
Synonym (2-Butyl-3-benzofuranyl)[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone hydrochloride; 2-Butyl-3-benzofuranyl-4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl ketone hydrochloride
Type Certified Reference Material
Quantity 1 g
Formula Weight 681.77
Grade Certified Reference Material