missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ aminoxyTMTsixplex™ Label Reagent Set

Catalog No. PI90402
Change view
Click to view available options
Quantity:
1 x 0.2 mg (per tag)
5 x 0.2 mg (per tag)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI90402 5 x 0.2 mg (per tag)
PI90401 1 x 0.2 mg (per tag)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI90402 Supplier Thermo Scientific™ Supplier No. LSG90402
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The carbonyl-reactive Thermo Scientific™ aminoxyTMT™ (Tandem Mass Tag™) Label Reagents enable multiplexed characterization and quantitation of carbonyl-containing biomolecules (carbohydrates, steroids, oxidized proteins) by mass spectrometry (MS).

The aminoxyTMTsixplex Label Reagents share an identical structure but contain different numbers of 13C and 15N isotopes in the mass reporter. The optimized labeling procedure can be completed in one hour. After tag labeling, samples are quenched and cleaned-up using HILIC solid-phase extraction before mass spectrometry analysis. The six compounds of the aminoxyTMTsixplex™ Reagent Set have the same mass (i.e., isobaric) and chemical structure (carbonyl-reactive aminoxy group, spacer arm, and mass reporter). However, the specific distribution of 13C and 15N isotopes on either side of the HCD or ETD MS/MS fragmentation site in each reagent results in a unique reporter mass (126-131Da) in the low mass region.

This set of reporter ions is used to measure the relative abundance of labeled molecules in a combined (multiplexed) MS sample representing six different treatment conditions. For glycobiology MS applications, the reagents enable quantitative profiling of glycan isoforms and discovery of glycan biomarkers; they provide improved ionization of labeled glycans for increased sensitivity and better retention of labeled glycans by reversed-phase liquid chromatography (RPLC). The aminoxyTMT reagents may be used to quantify a broad range of biologically important molecules including carbohydrates, steroids, or oxidized proteins.

Features:

  • Quantitative – enables relative quantitation of glycans or other carbonyl-containing biomolecules from multiple samples derived from cells, tissues or biological fluids
  • Stable – oxime bond and products formed by labeling reaction are stable and don't require an additional reduction step
  • Efficient – achieve labeling efficiency greater than 90% in one hour
  • Sensitive – 20-fold increase in signal-to-noise compared to unlabeled native glycan MS analysis
  • Multiplex – able to identify and characterize up to six samples concurrently
  • Optimized – procedure and reagents optimized for excellent labeling efficiency and recovery of glycans

Includes:

  • Label reagent sets contain sufficient aminoxyTMT tags for five sixplex experiments

Requires:

  • PNGase F (or PNGase A) glycosidase and Waters Oasis™ HLB columns

Applications:

  • Multiplex up to 6 different samples concurrently in a single experiment
  • Relative quantitation of glycans
  • Study of structural diversity of protein glycosylation
  • Study of glycosylation in cell signaling and regulation
  • Study of cancer progression, biomarker discovery and analysis of biotherapeutics

Native glycans are difficult to study by mass spectrometry because of their poor ionization efficiency. Quantitation of glycans is particularly challenging due to the lack of standards for all naturally occurring glycans and difficulties reproducibly quantifying multiple samples. The aminoxy group has better reactivity with carbonyls and better stability of the labeled product compared to hydrazides. Labeling with the aminoxyTMT reagents improves ionization of glycans, thus improving sensitivity, and enables relative quantitation of glycans for up to six samples concurrently.

These products are subject to a limited use label license.

Specifications

For Use With (Equipment) Mass Spectrometer
Product Type aminoxyTMTsixplex™ Label Reagent Set
Product Line aminoxyTMTsixplex
Content And Storage Sufficient for: Five sixplex (5 x 6-way) experiments
• aminoxyTMT6-126 Label Reagent, 5 x 0.2 mg
• aminoxyTMT6-127 Label Reagent, 5 x 0.2 mg
• aminoxyTMT6-128 Label Reagent, 5 x 0.2 mg
• aminoxyTMT6-129 Label Reagent, 5 x 0.2 mg
• aminoxyTMT6-130 Label Reagent, 5 x 0.2 mg
• aminoxyTMT6-131 Label Reagent, 5 x 0.2 mg

Store at –20°C.

Label or Dye Tandem Mass Tag™ (TMT) Labels
Starting Material Cell Lysate
Final Product Type Protein
Detection Method Mass Spectrometry
Workflow Step Protein Labeling
Quantity 5 x 0.2 mg (per tag)

For Research Use Only. Not for use in diagnostic procedures.