missing translation for 'onlineSavingsMsg'
Learn More

Aminophylline 98.0+%, TCI America™

Catalog No. p-7118086
Change view
Click to view available options
Quantity:
25 g
100 g
Catalog No. Quantity
A280525G 25 g
A2805100G 100 g
2 options

Catalog No. A280525G

Supplier: TCI America A280525G

Only null left

Chemical Identifiers

CAS 317-34-0
Molecular Formula C16H24N10O4
Molecular Weight (g/mol) 420.434
MDL Number MFCD00013221
InChI Key FQPFAHBPWDRTLU-UHFFFAOYSA-N
Synonym aminophylline, aminophyllin, theophyllamine, cardophyllin, phyllocontin, somophyllin, truphylline, theophylline ethylenediamine, cardiofilina, ethophylline
PubChem CID 9433
IUPAC Name 1,3-dimethyl-7H-purine-2,6-dione;ethane-1,2-diamine
SMILES CN1C2=C(C(=O)N(C1=O)C)NC=N2.CN1C2=C(C(=O)N(C1=O)C)NC=N2.C(CN)N

Specifications

Melting Point 272°C
Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 420.43
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Aminophylline
RTECSNumber XH5600000
TSCA Yes
Recommended Storage Freezer