missing translation for 'onlineSavingsMsg'
Learn More

Aminophylline, anhydrous, 98%

Catalog No. AAJ6070514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6070514 25 g
AAJ6070522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6070514 Supplier Thermo Scientific Chemicals Supplier No. J6070514

A non-selective phosphodiesterase inhibitor | CAS: 317-34-0 | C16H24N10O4 | 420.434 g/mol

Aminophylline exhibits vasodilatory and anti-inflammatory activity and is occasionally clinically used to treat chronic obstructive pulmonary disorder (COPD). In animal models of allergen-induced lung inflammation, aminophylline decreases levels of IL-5, IL-8, and eosinophils. It is mostly used to prepare solutions for intravenous injections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water. Insoluble in alcohol and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away fom strong oxidizing agents.

Chemical Identifiers

CAS 317-34-0
Molecular Formula C16H24N10O4
Molecular Weight (g/mol) 420.434
MDL Number MFCD00013221
InChI Key FQPFAHBPWDRTLU-UHFFFAOYSA-N
Synonym aminophylline, aminophyllin, theophyllamine, cardophyllin, phyllocontin, somophyllin, truphylline, theophylline ethylenediamine, cardiofilina, ethophylline
PubChem CID 9433
IUPAC Name 1,3-dimethyl-7H-purine-2,6-dione;ethane-1,2-diamine
SMILES CN1C2=C(C(=O)N(C1=O)C)NC=N2.CN1C2=C(C(=O)N(C1=O)C)NC=N2.C(CN)N

Specifications

Melting Point 269°C to 274°C
Color White to Yellow
Quantity 25 g
UN Number UN2811
Merck Index 14,465
Solubility Information Soluble in water. Insoluble in alcohol and ether.
Formula Weight 210.21
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Aminophylline, anhydrous
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 206-264-5
RTECSNumber XH5600000
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only