missing translation for 'onlineSavingsMsg'
Learn More

(Aminomethyl)phosphonic acid, 99%

CAS: 1066-51-9 | CH6NO3P | 111.037 g/mol

Supplier:  Thermo Scientific Chemicals L0983306

 View more versions of this product

Catalog No. AAL0983306


Only null left
Explore available promotions

Description

Description

(Aminomethyl)phosphonic acid is used in the preparation of (acetylamino-methyl)-phosphonic acid by reacting with acetic anhydride. Further, it is used in the development of immobilized metal affinity chromatography column based on organic-inorganic hybrid silica monolith.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Aminomethyl)phosphonic acid is used in the preparation of (acetylamino-methyl)-phosphonic acid by reacting with acetic anhydride. Further, it is used in the development of immobilized metal affinity chromatography column based on organic-inorganic hybrid silica monolith.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong bases and oxidizing agents.
Specifications

Specifications

(Aminomethyl)phosphonic acid
∽290°C (decomposition)
MFCD00008105
1701219
aminomethyl phosphonic acid, aminomethanephosphonic acid, amep, 1-aminomethylphosphonic acid, 1-aminomethylphosphonate, amino methane phosphoric acid, caswell no. 037c, 1-aminomethyl phosphonic acid, phosphonic acid, aminomethyl
MGRVRXRGTBOSHW-UHFFFAOYSA-N
aminomethylphosphonic acid
14017
111.04
1066-51-9
CH6NO3P
5 g
Hygroscopic
Soluble in water.
C(N)P(=O)(O)O
111.037
CHEBI:28812
99%

Chemical Identifiers

1066-51-9
111.037
MGRVRXRGTBOSHW-UHFFFAOYSA-N
14017
aminomethylphosphonic acid
CH6NO3P
MFCD00008105
aminomethyl phosphonic acid, aminomethanephosphonic acid, amep, 1-aminomethylphosphonic acid, 1-aminomethylphosphonate, amino methane phosphoric acid, caswell no. 037c, 1-aminomethyl phosphonic acid, phosphonic acid, aminomethyl
CHEBI:28812
C(N)P(=O)(O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 000-000-0

RTECSNumber : SZ6526000

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only