missing translation for 'onlineSavingsMsg'
Learn More

(Aminomethyl)phosphonic acid, 99%

Catalog No. p-7045206
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
250 mg
5 g
Catalog No. Quantity
AAL09833MD 250 mg
AAL0983303 1 g
AAL0983306 5 g
3 options

Catalog No. AAL09833MD

Supplier: Thermo Scientific Chemicals L09833MD

Only null left

CAS: 1066-51-9 | CH6NO3P | 111.037 g/mol

(Aminomethyl)phosphonic acid is used in the preparation of (acetylamino-methyl)-phosphonic acid by reacting with acetic anhydride. Further, it is used in the development of immobilized metal affinity chromatography column based on organic-inorganic hybrid silica monolith.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Aminomethyl)phosphonic acid is used in the preparation of (acetylamino-methyl)-phosphonic acid by reacting with acetic anhydride. Further, it is used in the development of immobilized metal affinity chromatography column based on organic-inorganic hybrid silica monolith.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong bases and oxidizing agents.

Chemical Identifiers

CAS 1066-51-9
Molecular Formula CH6NO3P
Molecular Weight (g/mol) 111.037
MDL Number MFCD00008105
InChI Key MGRVRXRGTBOSHW-UHFFFAOYSA-N
Synonym aminomethyl phosphonic acid, aminomethanephosphonic acid, amep, 1-aminomethylphosphonic acid, 1-aminomethylphosphonate, amino methane phosphoric acid, caswell no. 037c, 1-aminomethyl phosphonic acid, phosphonic acid, aminomethyl
PubChem CID 14017
ChEBI CHEBI:28812
IUPAC Name aminomethylphosphonic acid
SMILES C(N)P(=O)(O)O

Specifications

Melting Point ∽290°C (decomposition)
Quantity 250 mg
Beilstein 1701219
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 111.04
Percent Purity 99%
Chemical Name or Material (Aminomethyl)phosphonic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 000-000-0
RTECSNumber SZ6526000
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only