missing translation for 'onlineSavingsMsg'
Learn More
Learn More
δ-Aminolevulinic acid hydrochloride, 99.7%, MP Biomedicals™
delta-Aminolevulinic acid hydrochloride is the universal precursor of tetrapyrroles, such as chlorophyll and heme
$91.83 - $394.81
Chemical Identifiers
| CAS | 9-2-5451 |
|---|---|
| Molecular Formula | C5H10ClNO3 |
| Molecular Weight (g/mol) | 167.59 |
| MDL Number | MFCD00012869 |
| InChI Key | ZLHFONARZHCSET-UHFFFAOYSA-N |
| Synonym | 5-aminolevulinic acid hydrochloride, 5-amino-4-oxopentanoic acid hydrochloride, aminolevulinic acid hydrochloride, levulan, levulan kerastick, aminolevulinic acid hcl, delta-aminolevulinic acid hydrochloride, gliolan, ameluz, 5-aminolevulinate hydrochloride |
| PubChem CID | 123608 |
| IUPAC Name | 5-amino-4-oxopentanoic acid;hydrochloride |
| SMILES | C(CC(=O)O)C(=O)CN.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN10024790
|
MP Biomedicals Inc
0210024790 |
500 mg |
N/A
|
|
|||||
|
ICN10024780
|
MP Biomedicals Inc
0210024780 |
0.1 g |
N/A
|
|
|||||
|
ICN10024791
|
MP Biomedicals Inc
0210024791 |
1 g |
N/A
|
|
|||||
Chemical Identifiers
| 9-2-5451 | |
| 167.59 | |
| ZLHFONARZHCSET-UHFFFAOYSA-N | |
| 123608 | |
| C(CC(=O)O)C(=O)CN.Cl |
| C5H10ClNO3 | |
| MFCD00012869 | |
| 5-aminolevulinic acid hydrochloride, 5-amino-4-oxopentanoic acid hydrochloride, aminolevulinic acid hydrochloride, levulan, levulan kerastick, aminolevulinic acid hcl, delta-aminolevulinic acid hydrochloride, gliolan, ameluz, 5-aminolevulinate hydrochloride | |
| 5-amino-4-oxopentanoic acid;hydrochloride |
Specifications
| 9-2-5451 | |
| 0.997 | |
| MFCD00012869 | |
| ZLHFONARZHCSET-UHFFFAOYSA-N | |
| 5-amino-4-oxopentanoic acid;hydrochloride | |
| 167.59 | |
| 167.6 | |
| δ-Aminolevulinic acid hydrochloride |
| White | |
| C5H10ClNO3 | |
| 5-aminolevulinic acid hydrochloride, 5-amino-4-oxopentanoic acid hydrochloride, aminolevulinic acid hydrochloride, levulan, levulan kerastick, aminolevulinic acid hcl, delta-aminolevulinic acid hydrochloride, gliolan, ameluz, 5-aminolevulinate hydrochloride | |
| C(CC(=O)O)C(=O)CN.Cl | |
| 500 mg | |
| 123608 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : 226-679-5