missing translation for 'onlineSavingsMsg'
Learn More
Learn More
δ-Aminolevulinic acid hydrochloride, 99.7%, MP Biomedicals™
delta-Aminolevulinic acid hydrochloride is the universal precursor of tetrapyrroles, such as chlorophyll and heme
Supplier: MP Biomedicals Inc 0210024780
Specifications
| δ-Aminolevulinic acid hydrochloride | |
| White | |
| C5H10ClNO3 | |
| 5-aminolevulinic acid hydrochloride, 5-amino-4-oxopentanoic acid hydrochloride, aminolevulinic acid hydrochloride, levulan, levulan kerastick, aminolevulinic acid hcl, delta-aminolevulinic acid hydrochloride, gliolan, ameluz, 5-aminolevulinate hydrochloride | |
| C(CC(=O)O)C(=O)CN.Cl | |
| 0.1 g | |
| 123608 | |
| Crystalline Powder |
| 9-2-5451 | |
| 0.997 | |
| MFCD00012869 | |
| ZLHFONARZHCSET-UHFFFAOYSA-N | |
| 5-amino-4-oxopentanoic acid;hydrochloride | |
| 167.59 | |
| 167.6 |
Chemical Identifiers
| 9-2-5451 | |
| 167.59 | |
| ZLHFONARZHCSET-UHFFFAOYSA-N | |
| 123608 | |
| C(CC(=O)O)C(=O)CN.Cl |
| C5H10ClNO3 | |
| MFCD00012869 | |
| 5-aminolevulinic acid hydrochloride, 5-amino-4-oxopentanoic acid hydrochloride, aminolevulinic acid hydrochloride, levulan, levulan kerastick, aminolevulinic acid hcl, delta-aminolevulinic acid hydrochloride, gliolan, ameluz, 5-aminolevulinate hydrochloride | |
| 5-amino-4-oxopentanoic acid;hydrochloride |
Safety and Handling
EINECSNumber : 226-679-5