missing translation for 'onlineSavingsMsg'
Learn More

Aminoguanidine Sulfate Hydrate 98.0+%, TCI America™

$54.06 - $425.00

Chemical Identifiers

CAS 996-19-0
Molecular Formula CH6N4
MDL Number MFCD00035665
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A030925G
View Documents
TCI America
A030925G
25 g
Each for $54.06
Only null left
 
A0309500G
View Documents
TCI America
A0309500G
500 g
Each for $425.00
Only null left
 
Description

Description

Specifications

Chemical Identifiers

996-19-0
CH6N4
MFCD00035665

Specifications

996-19-0
White
MFCD00035665
246.25
Crystalline Powder
207°C
CH6N4
25 g
≥98.0% (T)
Aminoguanidine Sulfate Hydrate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1776&(9)-0070

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'