missing translation for 'onlineSavingsMsg'
Learn More

Aminoguanidine Sulfate Hydrate 98.0+%, TCI America™

Supplier:  TCI America A030925G

 View more versions of this product

Catalog No. A030925G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Aminoguanidine Sulfate Hydrate
207°C
CH6N4
25 g
≥98.0% (T)
996-19-0
White
MFCD00035665
246.25
Crystalline Powder

Chemical Identifiers

996-19-0
CH6N4
MFCD00035665
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1776&(9)-0070

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions