missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminoferrocene, ≥98.0% (T); 98.0-102.0% (NT), MilliporeSigma™ Supelco™
$537.10 - $2146.26
Chemical Identifiers
| MDL Number | MFCD03844760 |
|---|---|
| Synonym | eta5-(1-Aminocyclopentadienyl)(cyclopentadienyl)iron(II); Fc-NH2; Ferrocenylamine |
Chemical Identifiers
| MFCD03844760 | |
| eta5-(1-Aminocyclopentadienyl)(cyclopentadienyl)iron(II); Fc-NH2; Ferrocenylamine |
Specifications
| 1273-82-1 | |
| MFCD03844760 | |
| eta5-(1-Aminocyclopentadienyl)(cyclopentadienyl)iron(II); Fc-NH2; Ferrocenylamine | |
| ≥98.0% (T); 98.0 to 102.0% (NT) |
| C10H11 FeN | |
| 250 mg | |
| 201.05 | |
| For HPLC derivatization |