missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminoferrocene, ≥98.0% (T); 98.0-102.0% (NT), MilliporeSigma™ Supelco™
Supplier: Merck Emd Millipore 073791G
Specifications
| 1273-82-1 | |
| MFCD03844760 | |
| eta5-(1-Aminocyclopentadienyl)(cyclopentadienyl)iron(II); Fc-NH2; Ferrocenylamine | |
| ≥98.0% (T); 98.0 to 102.0% (NT) |
| C10H11 FeN | |
| 1 g | |
| 201.05 | |
| For HPLC derivatization |
Chemical Identifiers
| MFCD03844760 | |
| eta5-(1-Aminocyclopentadienyl)(cyclopentadienyl)iron(II); Fc-NH2; Ferrocenylamine |