missing translation for 'onlineSavingsMsg'
Learn More

Aminodiphenylmethane, 97%

CAS: 91-00-9 | C13H13N | 183.25 g/mol

$144.77 - $144.77

Chemical Identifiers

CAS 91-00-9
Molecular Formula C13H13N
Molecular Weight (g/mol) 183.25
MDL Number MFCD00008059
InChI Key MGHPNCMVUAKAIE-UHFFFAOYSA-N
Synonym aminodiphenylmethane, benzhydrylamine, 1,1-diphenylmethylamine, diphenylmethyl amine, 1,1-diphenylmethanamine, alpha-aminodiphenylmethane, methanamine, 1,1-diphenyl, alpha-phenylbenzylamine, benzhydrylamin, benzhydryl amine
PubChem CID 7036
IUPAC Name diphenylmethanamine
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC304320250
SDS View Documents
Thermo Scientific Chemicals
304320250
25 g Glass bottle
Each for $144.77
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

91-00-9
183.25
MGHPNCMVUAKAIE-UHFFFAOYSA-N
7036
C1=CC=C(C=C1)C(C2=CC=CC=C2)N
C13H13N
MFCD00008059
aminodiphenylmethane, benzhydrylamine, 1,1-diphenylmethylamine, diphenylmethyl amine, 1,1-diphenylmethanamine, alpha-aminodiphenylmethane, methanamine, 1,1-diphenyl, alpha-phenylbenzylamine, benzhydrylamin, benzhydryl amine
diphenylmethanamine

Specifications

91-00-9
Colorless to Yellow
295.0°C
Authentic
Glass bottle
1.5950 to 1.5970
MFCD00008059
12, 1323
15, 1078
MGHPNCMVUAKAIE-UHFFFAOYSA-N
diphenylmethanamine
7036
97%
Aminodiphenylmethane
12.0°C
1.0600g/mL
>110°C
96% min. (GC)
C13H13N
(C6H5)2CHNH2
25 g
1.06
aminodiphenylmethane, benzhydrylamine, 1,1-diphenylmethylamine, diphenylmethyl amine, 1,1-diphenylmethanamine, alpha-aminodiphenylmethane, methanamine, 1,1-diphenyl, alpha-phenylbenzylamine, benzhydrylamin, benzhydryl amine
C1=CC=C(C=C1)C(C2=CC=CC=C2)N
183.25
183.25
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Warning

EINECSNumber : 202-032-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only