missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminocyclobutane 97.0+%, TCI America™
Supplier: TCI America A10365ML
Specifications
| Aminocyclobutane | |
| Yellow | |
| C4H9N | |
| 5 mL | |
| cyclobutylamine, aminocyclobutane, cyclobutyl amine, cyclobutylamin, cyclobutaneamine, 1-amino cyclobutane, pubchem11047, acmc-209ghn, cyclobutylammonium chloride | |
| C1CC(C1)N | |
| 71.123 | |
| 71.12 | |
| Liquid |
| 2516-34-9 | |
| 82°C | |
| MFCD00001328 | |
| 2733 | |
| KZZKOVLJUKWSKX-UHFFFAOYSA-N | |
| cyclobutanamine | |
| 75645 | |
| ≥97.0% (GC,T) |
Chemical Identifiers
| 2516-34-9 | |
| 71.123 | |
| KZZKOVLJUKWSKX-UHFFFAOYSA-N | |
| 75645 | |
| C1CC(C1)N |
| C4H9N | |
| MFCD00001328 | |
| cyclobutylamine, aminocyclobutane, cyclobutyl amine, cyclobutylamin, cyclobutaneamine, 1-amino cyclobutane, pubchem11047, acmc-209ghn, cyclobutylammonium chloride | |
| cyclobutanamine |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator