missing translation for 'onlineSavingsMsg'
Learn More

Aminocyclobutane 97.0+%, TCI America™

Catalog No. p-7117204
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10361ML 1 mL
A10365ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A10361ML Supplier TCI America Supplier No. A10361ML
Only null left

Chemical Identifiers

CAS 2516-34-9
Molecular Formula C4H9N
Molecular Weight (g/mol) 71.123
MDL Number MFCD00001328
InChI Key KZZKOVLJUKWSKX-UHFFFAOYSA-N
Synonym cyclobutylamine, aminocyclobutane, cyclobutyl amine, cyclobutylamin, cyclobutaneamine, 1-amino cyclobutane, pubchem11047, acmc-209ghn, cyclobutylammonium chloride
PubChem CID 75645
IUPAC Name cyclobutanamine
SMILES C1CC(C1)N

Specifications

Color Yellow
Boiling Point 82°C
Quantity 1 mL
UN Number 2733
Formula Weight 71.12
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Aminocyclobutane
TSCA No
Recommended Storage Refrigerator