missing translation for 'onlineSavingsMsg'
Learn More

Aminoacetal 97.0+%, TCI America™

Catalog No. p-7116788
Change view
Click to view available options
Quantity:
25 mL
250 mL
Catalog No. Quantity
A024725ML 25 mL
A0247250ML 250 mL
2 options

Catalog No. A024725ML

Supplier: TCI America A024725ML

Only null left

Chemical Identifiers

CAS 645-36-3
Molecular Formula C6H15NO2
Molecular Weight (g/mol) 133.19
MDL Number MFCD00008136
InChI Key HJKLEAOXCZIMPI-UHFFFAOYSA-N
Synonym 2,2-diethoxyethylamine, aminoacetaldehyde diethyl acetal, ethanamine, 2,2-diethoxy, 2,2-diethoxyethan-1-amine, 2-aminoacetaldehyde diethyl acetal, glycinaldehyde diethyl acetal, aminoacetal, diethoxyethylamine, acetaldehyde, amino-, diethyl acetal, aminoacetaldehydediethylacetal
PubChem CID 69524
IUPAC Name 2,2-diethoxyethan-1-amine
SMILES CCOC(CN)OCC

Specifications

Color Yellow
Boiling Point 163°C
Quantity 25 mL
UN Number 2924
Formula Weight 133.19
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Aminoacetal