missing translation for 'onlineSavingsMsg'
Learn More

Amino Acid Standards, Physiological, Analytical Standard, Acidics and Neutrals, MilliporeSigma™ Supelco™

Catalog No. 111017237
Change view
Click to view available options
Quantity:
1 mL
5 mL
10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017237 1 mL
111017236 5 mL
111017235 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111017237 Supplier Merck Emd Millipore Supplier No. A64071ML

Amino acids listed are 2.5 mumoles per mL except L-cystine at 1.25 mumoles per ml.

Specifications

Packaging Glass Bottle
Physical Form Neat
MDL Number MFCD00162052
Quantity 1 mL
Grade Analytical Standard