missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amino Acid Standards, For Protein Hydrolysates containing norleucine, Analytical Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| Packaging | Glass Bottle |
| Physical Form | Neat |
| Quantity | 5 mL |
| Grade | Analytical Standard |