Learn More
Amine-Sulfuric Acid Reagent, For Sulfide, Certified, LabChem™
Supplier: LabChem LC108802
Specifications
| Amine-Sulfuric Acid Reagent | |
| For Sulfide | |
| 58.91,40.41,0.68 | |
| C18H26N4O4 | |
| UN1830 | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| sulfuric acid | |
| 1118 | |
| 181.21 | |
| Passes Test | |
| 1.49g/mL | |
| Sulfur compounds; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors; Nitrogen oxides | |
| Liquid |
| Amber to Pink | |
| 7664-93-9,7732-18-5,62778-12-5 | |
| H2O4S | |
| MFCD00064589 | |
| 1.49g/mL | |
| Soluble in water | |
| OS(O)(=O)=O | |
| 98.07 | |
| CHEBI:26836 | |
| Certified | |
| Amber Glass | |
| Passes Test | |
| 1 L |
Chemical Identifiers
| 7664-93-9 | |
| 98.07 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| 1118 | |
| sulfuric acid |
| H2O4S | |
| MFCD00064589 | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| CHEBI:26836 | |
| OS(O)(=O)=O |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life.
GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature