missing translation for 'onlineSavingsMsg'
Learn More

Amiloride hydrochloride dihydrate

Catalog No. p-7058213
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6216803 1 g
AAJ6216806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6216803 Supplier Thermo Scientific Chemicals Supplier No. J6216803
Only null left

Sodium channel blocker | CAS: 17440-83-4 | C6H13Cl2N7O3 | 302.116 g/mol

Amiloride hydrochloride dihydrate is used as a calcium channel and sodium channel protein inhibitor. It reduces electrical potential across tubular epithelium, inhibits angiogenesis and inhibits capillary morphogenesis completely and reversibly at approximately 130 ?M. Amiloride

  • HCl is an inhibitor of Sodium/Potassium-ATPase Protein. Defines the I2A-amiloride sensitive and I2B-amiloride insensitive imidazoline binding Blocks TRPP3, acid sensing- (ASIC) and mechanogated membrane-ion channels, as well as the Na+/H+ exchanger. Also inhibits urokinase-type plasminogen activator (uPA); has no effect on tissue-type plasminogen activator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amiloride hydrochloride dihydrate is used as a calcium channel and sodium channel protein inhibitor. It reduces electrical potential across tubular epithelium, inhibits angiogenesis and inhibits capillary morphogenesis completely and reversibly at approximately 130 μM. Amiloride • HCl is an inhibitor of Sodium/Potassium-ATPase Protein. Defines the I2A-amiloride sensitive and I2B-amiloride insensitive imidazoline binding Blocks TRPP3, acid sensing- (ASIC) and mechanogated membrane-ion channels, as well as the Na+/H+ exchanger. Also inhibits urokinase-type plasminogen activator (uPA); has no effect on tissue-type plasminogen activator.

Solubility
Soluble in water at 50mg/ml

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 17440-83-4
Molecular Formula C6H13Cl2N7O3
Molecular Weight (g/mol) 302.116
MDL Number MFCD00058197
InChI Key LTKVFMLMEYCWMK-UHFFFAOYSA-N
Synonym amiloride hydrochloride dihydrate, modamide, nirulid, amiloride hcl, amiloride hcl dihydrate, amilorid hydrochlorid-2-wasser, unii-fzj37245uc, amiloride hydrochloride usan:usp, amiloride dihydrate hydrochloride, n-amidino-3,5-diamino-6-chloropyrazinecarboxamide hydrochloride dihydrate
PubChem CID 68540
ChEBI CHEBI:2640
IUPAC Name 3,5-diamino-6-chloro-N-(diaminomethylidene)pyrazine-2-carboxamide;dihydrate;hydrochloride
SMILES C1(=C(N=C(C(=N1)Cl)N)N)C(=O)N=C(N)N.O.O.Cl

Specifications

Melting Point >240°C
Color Yellow
Quantity 1 g
UN Number UN2811
Merck Index 14,406
Solubility Information Soluble in water at 50mg/ml
Formula Weight 266.09
Physical Form Powder
Chemical Name or Material Amiloride hydrochloride dihydrate
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 217-958-2
RTECSNumber UQ2275500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only