missing translation for 'onlineSavingsMsg'
Learn More

Amikacin disulfate salt, Thermo Scientific Chemicals

$194.24 - $194.24

Chemical Identifiers

CAS 39831-55-5
Molecular Formula C22H47N5O21S2
Molecular Weight (g/mol) 781.75
MDL Number MFCD00167475,MFCD00167475
InChI Key FXKSEJFHKVNEFI-GCZBSULCSA-N
PubChem CID 134129615
IUPAC Name (2S)-4-amino-N-[(1R,2S,3S,4S,5S)-5-amino-2-[(2S,3S,4S,5S,6S)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4-[(2R,3S,4S,5S,6S)-6-(aminomethyl)-3,4,5-trihydroxyoxan-2-yl]oxy-3-hydroxycyclohexyl]-2-hydroxybutanamide;sulfuric acid
SMILES OS(O)(=O)=O.OS(O)(=O)=O.NCC[C@H](O)C(=O)N[C@@H]1C[C@H](N)[C@@H](O[C@H]2O[C@H](CN)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O[C@H]1O[C@H](CO)[C@@H](O)[C@H](N)[C@H]1O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC455190050
View Documents
Thermo Scientific Chemicals
455190050
5 g
Each for $194.24
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

39831-55-5
781.75
FXKSEJFHKVNEFI-GCZBSULCSA-N
(2S)-4-amino-N-[(1R,2S,3S,4S,5S)-5-amino-2-[(2S,3S,4S,5S,6S)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4-[(2R,3S,4S,5S,6S)-6-(aminomethyl)-3,4,5-trihydroxyoxan-2-yl]oxy-3-hydroxycyclohexyl]-2-hydroxybutanamide;sulfuric acid
C22H47N5O21S2
MFCD00167475,MFCD00167475
134129615
OS(O)(=O)=O.OS(O)(=O)=O.NCC[C@H](O)C(=O)N[C@@H]1C[C@H](N)[C@@H](O[C@H]2O[C@H](CN)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O[C@H]1O[C@H](CO)[C@@H](O)[C@H](N)[C@H]1O

Specifications

39831-55-5
2 to 4 (1% aq. soln.)
Authentic
MFCD00167475,MFCD00167475
Solubility in water: ca 50g/L.
OS(O)(=O)=O.OS(O)(=O)=O.NCC[C@H](O)C(=O)N[C@@H]1C[C@H](N)[C@@H](O[C@H]2O[C@H](CN)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O[C@H]1O[C@H](CO)[C@@H](O)[C@H](N)[C@H]1O
781.75
781.76
220.0°C to 230.0°C
13% max. (110°C, 3 h, 5 mm Hg)
C22H47N5O21S2
5 g
FXKSEJFHKVNEFI-GCZBSULCSA-N
(2S)-4-amino-N-[(1R,2S,3S,4S,5S)-5-amino-2-[(2S,3S,4S,5S,6S)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4-[(2R,3S,4S,5S,6S)-6-(aminomethyl)-3,4,5-trihydroxyoxan-2-yl]oxy-3-hydroxycyclohexyl]-2-hydroxybutanamide;sulfuric acid
134129615
Amikacin disulfate salt
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 254-648-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only