missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMIGO1 Mouse anti-Human, Rat, FITC, Clone:S86-36, Abnova™
Mouse monoclonal antibody raised against partial recombinant human AMIGO1.
Supplier: Abnova Corporation MAB17351
Specifications
| AMIGO1 | |
| Monoclonal | |
| FITC | |
| Immunocytochemistry Immunofluorescence Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Western Blot (1:1000) The optimal working dilution should be determined by the end user. |
|
| AMIGO1 | |
| Mouse | |
| 100 μg | |
| Human, Rat | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| S86-36 | |
| adhesion molecule with Ig-like domain 1 | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
| AMIGO | |
| Recombinant protein corresponding to amino acids 554-574 at C-terminus of human AMIGO1. | |
| 57463 | |
| Store at -20°C. | |
| Liquid |