missing translation for 'onlineSavingsMsg'
Learn More

Amidosulfonic acid, 99.99% (metals basis)

Catalog No. p-7059396
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4451014 25 g
AA4451022 100 g
AA4451036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4451014 Supplier Thermo Scientific Chemicals Supplier No. 04451014
Only null left

CAS: 5329-14-6 | H3NO3S | 97.088 g/mol

Amidosulfonic acid is mainly a precursor to sweet-tasting compounds. Reaction with cyclohexylamine followed by addition of NaOH gives C6H11NHSO3Na, sodium cyclamate. Sulfamic acid is used as an acidic cleaning agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5329-14-6
Molecular Formula H3NO3S
Molecular Weight (g/mol) 97.088
MDL Number MFCD00011603
InChI Key IIACRCGMVDHOTQ-UHFFFAOYSA-N
Synonym amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid
PubChem CID 5987
ChEBI CHEBI:9330
IUPAC Name sulfamic acid
SMILES NS(=O)(=O)O

Specifications

pH 1.2
Melting Point ∼205°C (decomposition)
Density 2.12 g/mL
Odor Odorless
Quantity 25 g
Physical Form Crystalline
UN Number UN2967
Merck Index 148921
Solubility Information Soluble in water.
Formula Weight 97.09
Percent Purity 99.99%
Assay Metals basis
Chemical Name or Material Amidosulfonic acid
Show More Show Less
Hazard Category H290-H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P234-P264b-P280g-P280i-P302+P352-P305+P351+P338-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: SULFAMIC ACID
EINECSNumber 226-218-8
RTECSNumber W05950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only