missing translation for 'onlineSavingsMsg'
Learn More

Amidol 97.0+%, TCI America™

Catalog No. p-7126016
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D008025G 25 g
D0080500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D008025G Supplier TCI America Supplier No. D008025G
Only null left

Chemical Identifiers

CAS 137-09-7
Molecular Formula C6H10Cl2N2O
Molecular Weight (g/mol) 197.06
MDL Number MFCD00012979
InChI Key KQEIJFWAXDQUPR-UHFFFAOYSA-N
Synonym 2,4-diaminophenol dihydrochloride, amidol, dianol, acrol, 2,4-diaminophenol hcl, 2,4-diaminophenol.2hcl, phenol, 2,4-diamino-, dihydrochloride, 2,4-diaminophenol hydrochloride, diamidophenol hydrochloride, unii-t1iyy101nx
PubChem CID 8715
IUPAC Name dihydrogen 2,4-diaminophenol dichloride
SMILES [H+].[H+].[Cl-].[Cl-].NC1=CC=C(O)C(N)=C1

Specifications

Melting Point 205°C
Color Green-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 197.06
Percent Purity ≥97.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Amidol
EINECSNumber (3)-2772
RTECSNumber SK7600000
TSCA Yes