missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amidol 98.0+%, TCI America™
$115.57 - $115.57
Chemical Identifiers
| CAS | 137-09-7 |
|---|---|
| Molecular Formula | C6H10Cl2N2O |
| Molecular Weight (g/mol) | 197.06 |
| MDL Number | MFCD00012979 |
| InChI Key | KQEIJFWAXDQUPR-UHFFFAOYSA-N |
| Synonym | 2,4-diaminophenol dihydrochloride, amidol, dianol, acrol, 2,4-diaminophenol hcl, 2,4-diaminophenol.2hcl, phenol, 2,4-diamino-, dihydrochloride, 2,4-diaminophenol hydrochloride, diamidophenol hydrochloride, unii-t1iyy101nx |
| PubChem CID | 8715 |
| IUPAC Name | dihydrogen 2,4-diaminophenol dichloride |
| SMILES | [H+].[H+].[Cl-].[Cl-].NC1=CC=C(O)C(N)=C1 |
Chemical Identifiers
| 137-09-7 | |
| 197.06 | |
| KQEIJFWAXDQUPR-UHFFFAOYSA-N | |
| 8715 | |
| [H+].[H+].[Cl-].[Cl-].NC1=CC=C(O)C(N)=C1 |
| C6H10Cl2N2O | |
| MFCD00012979 | |
| 2,4-diaminophenol dihydrochloride, amidol, dianol, acrol, 2,4-diaminophenol hcl, 2,4-diaminophenol.2hcl, phenol, 2,4-diamino-, dihydrochloride, 2,4-diaminophenol hydrochloride, diamidophenol hydrochloride, unii-t1iyy101nx | |
| dihydrogen 2,4-diaminophenol dichloride |
Specifications
| 137-09-7 | |
| C6H10Cl2N2O | |
| 25 g | |
| 2,4-diaminophenol dihydrochloride, amidol, dianol, acrol, 2,4-diaminophenol hcl, 2,4-diaminophenol.2hcl, phenol, 2,4-diamino-, dihydrochloride, 2,4-diaminophenol hydrochloride, diamidophenol hydrochloride, unii-t1iyy101nx | |
| [H+].[H+].[Cl-].[Cl-].NC1=CC=C(O)C(N)=C1 | |
| 197.06 | |
| 197.06 | |
| Crystalline Powder |
| 205°C | |
| MFCD00012979 | |
| 2811 | |
| KQEIJFWAXDQUPR-UHFFFAOYSA-N | |
| dihydrogen 2,4-diaminophenol dichloride | |
| 8715 | |
| ≥98.0% (N,T) | |
| Amidol |
Safety and Handling
EINECSNumber : (3)-2772
RTECSNumber : SK7600000
TSCA : Yes