missing translation for 'onlineSavingsMsg'
Learn More

Amidinothiourea, 99+%

CAS: 2114-02-5 | C2H6N4S | 118.16 g/mol

$59.94 - $59.94

Chemical Identifiers

CAS 2114-02-5
Molecular Formula C2H6N4S
Molecular Weight (g/mol) 118.16
MDL Number MFCD00014472
InChI Key OKGXJRGLYVRVNE-UHFFFAOYSA-N
Synonym amidinothiourea, guanylthiourea, guthimin, guthimine, 2-imino-4-thiobiuret, n-amidinothiourea, thiourea, aminoiminomethyl, thiodicyanodiamidine, diaminomethylidene thiourea, gutimine
PubChem CID 2724563
IUPAC Name diaminomethylidenethiourea
SMILES C(=NC(=S)N)(N)N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC259190100
SDS View Documents
Thermo Scientific Chemicals
259190100
10 g Glass bottle
Each for $59.94
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2114-02-5
118.16
OKGXJRGLYVRVNE-UHFFFAOYSA-N
2724563
C(=NC(=S)N)(N)N
C2H6N4S
MFCD00014472
amidinothiourea, guanylthiourea, guthimin, guthimine, 2-imino-4-thiobiuret, n-amidinothiourea, thiourea, aminoiminomethyl, thiodicyanodiamidine, diaminomethylidene thiourea, gutimine
diaminomethylidenethiourea

Specifications

2114-02-5
White
Authentic
Glass bottle
NH2C(=NH)NHC(=S)NH2
10 g
Solubility in water: 7 w/w % in water (20°C)
C(=NC(=S)N)(N)N
118.16
118.16
Crystalline Powder or Crystals
160°C to 164°C
0.3% max.
99% min. (Iodimetry)
C2H6N4S
MFCD00014472
amidinothiourea, guanylthiourea, guthimin, guthimine, 2-imino-4-thiobiuret, n-amidinothiourea, thiourea, aminoiminomethyl, thiodicyanodiamidine, diaminomethylidene thiourea, gutimine
OKGXJRGLYVRVNE-UHFFFAOYSA-N
diaminomethylidenethiourea
2724563
99+%
Amidinothiourea, 99+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 218-308-0

RTECSNumber : YR8250000

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only