missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Amicon™ Stirred Cells

Catalog No. UFSC40001
Change view
Click to view available options
Volume (Metric) Hold Up:
50 mL
200 mL
400 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Hold Up
UFSC40001 400 mL
UFSC05001 50 mL
UFSC20001 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Pressure Limitation. Do not operate the stirred cell above 5.2 bar (75 psi).
Catalog No. UFSC40001 Supplier MilliporeSigma™ Supplier No. UFSC40001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pressure Limitation. Do not operate the stirred cell above 5.2 bar (75 psi).

Amicon stirred cells concentrate samples rapidly but gently, using magnetic stirring to minimize concentration polarization and shear stress-induced denaturation.

This Amicon stirred cell model provides high flow rates with solutions up to 10% macrosolute concentration. Capable of rapid concentration, or salt removal followed by concentration in the same unit.

Warranty

One year

Specifications

Autoclavable Autoclavable
Description Amicon Stirred Cell 400 mL
For Use With (Application) Concentration, Buffer Exchange, Desalting
Includes Amicon™ Stirred Cell 400 mL
Type Ultrafiltration Equipment
Volume (Metric) Hold Up 400 mL
For Use With (Equipment) Ultracel™, Biomax™, and Durapore™ membrane discs for use in stirred cells