missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Amicon™ Stirred Cell Maintenance Kits (Stir Bar Assembly, 2 Seals, Tubing kit)

Amicon stirred cells concentrate samples rapidly but gently, using magnetic stirring to minimize concentration polarization and shear stress-induced denaturation.

Supplier:  MilliporeSigma™ UFSC200MT

 View more versions of this product

Catalog No. UFSC200MT


Only null left

Description

Description

Amicon stirred cells concentrate samples rapidly but gently, using magnetic stirring to minimize concentration polarization and shear stress-induced denaturation.

This Amicon stirred cell model provides high flow rates with solutions up to 10% macrosolute concentration. Capable of rapid concentration, or salt removal followed by concentration in the same unit.
Specifications

Chemical Identifiers

Ultrafiltration Equipment

Specifications

Amicon Stirred Cell 200 mL Maintenance kit
Amicon Stirred Cell (200 mL)
Amicon™ Stirred Cell 200 mL Maintenance kit (stir bar assembly, 2 Ea. seals, tubing kit)
Ultrafiltration Equipment
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions