missing translation for 'onlineSavingsMsg'
Learn More

Ametycin 98.0+%, TCI America™

Catalog No. M232010MG
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M232010MG 10 mg
M232050MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M232010MG Supplier TCI America Supplier No. M232010MG

Chemical Identifiers

CAS 50-07-7
Molecular Formula C15H18N4O5
Molecular Weight (g/mol) 334.332
MDL Number MFCD00078109
InChI Key NWIBSHFKIJFRCO-WUDYKRTCSA-N
Synonym mitomycin c, mitomycin, ametycine, mutamycin, ametycin, mitocin-c, mitomycinum, mytomycin, mytozytrex, mitomycin-c
PubChem CID 5746
ChEBI CHEBI:27504
SMILES CC1=C(C(=O)C2=C(C1=O)N3CC4C(C3(C2COC(=O)N)OC)N4)N

Specifications

Melting Point 64°C
Color Blue-Purple
Quantity 10 mg
UN Number 2811
Formula Weight 334.33
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Ametycin
RTECSNumber CN0700000
TSCA Yes
Recommended Storage Refrigerator