Learn More
Cytiva Amersham™ Reinforced Nitrocellulose Blotting Membrane
Nitrocellulose membrane, with a 0.45 μm pore size, is suitable for proteins of a wide range of molecular weights.
Supplier: Cytiva 10600124
Description
- Low background and excellent signal-to-noise ratios.
- Compatible with a variety of labeling and detection systems.
- High binding capacity.
- Suitable for proteins of a wide range of molecular weights.
- Proprietary nitrocellulose formula gives a long shelf life of bound proteins.
- Available in popular precut sizes for your application.
Specifications
| Blotting Membrane | |
| Protein | |
| 1 Each | |
| For western, southern and northern blotting applications |