missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ ECL™ Western Blotting Detection Reagents

Catalog No. 45000885
Change view
Click to view available options
Quantity:
62.5 mL
125 mL
250 mL
375 mL
Sufficient For:
1000 cm2 Membrane
2000 cm2 Membrane
4000 cm2 Membrane
6000 cm2 Membrane
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sufficient For
45000885 125 mL 2000 cm2 Membrane
45000878 62.5 mL 1000 cm2 Membrane
45001160 375 mL 6000 cm2 Membrane
45000875 250 mL 4000 cm2 Membrane
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 45000885 Supplier Cytiva Supplier No. RPN2209
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use for all routine Western blotting. Cytiva Life Sciences™ Amersham™ ECL™ Western Blotting Detection Reagents are the world's first commercially available chemiluminescent detection reagents and still one of the most widely used.

  • Detectable on Hyperfilm™ ECL or with CCD imager
  • Uses enhanced luminol-based detection which is suitable for all routine Western Blotting

Western Blotting

Specifications

Product Type ECL Western Blot Detection Reagent
For Use With (Application) Western Blot
Quantity 125 mL
Sensitivity ∼10 pg
Sufficient For 2000 cm2 Membrane
Content And Storage 125mL detection reagent, 1125mL detection reagent 2 2°C to 8°C