missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ ECL™ start Western Blotting Detection Reagent

Catalog No. 45100100
Change view
Click to view available options
Quantity:
200 mL
400 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45100100 200 mL
45100101 400 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45100100 Supplier Cytiva Supplier No. RPN3243
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Reagent is luminol-based chemiluminescent substrate designed for the detection of proteins immobilized to a membrane

Amersham™ ECL™ start is a chemiluminescent detection reagent in the Amersham ECL product family that is recommended for confirmatory analysis and detection of high to medium levels of endogenous proteins. Amersham ECL start has a long signal duration, which enables multiple exposures of the membrane plus a convenient time window between experiment and analysis.

  • Long signal duration provides assay design flexibility and robust detection of large experimental sets
  • Produces low background, regardless of method of detection
  • Working solution is stable for five days
  • Long shelf life of 18 hours is convenient for those who run Western blots infrequently
  • Detection on X-ray film or CCD Imager
  • Recommended Membrane: PVDF or nitrocellulose
  • For detection of high abundant proteins
  • Colorless detection reagent 1 and 2

Applications:

Routine and confirmatory Western blotting

Specifications

Product Type ECL Western Blotting Detection Reagent
Quantity 200 mL
For Use With (Application) Routine and confirmatory Western blotting
Storage Requirements 18°C to 30°C