missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ ECL™ Prime Western Blotting Detection Reagent
GREENER_CHOICE

Catalog No. 45002401
Change view
Click to view available options
Sufficient For:
1000cm2 membrane
3000cm2 membrane
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
45002401 1000cm2 membrane
45010090 3000cm2 membrane
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45002401 Supplier Cytiva Supplier No. RPN2232
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Solution A (luminol solution) and Solution B (peroxide solution)

Enables the use of highly diluted primary and secondary antibodies without reducing sensitivity. Cytiva Lifescience™ Amersham™ ECL™ Prime Western Blotting Detection Reagent produces high signal intensity and sensitivity.

  • Stable signal enables multiple exposures and makes reagent suitable for large experimental series
  • Provides convenient handling time between detection and completion of experiment
  • Optimized for imaging with ImageQuant™ LAS 4000 (CCD-based imaging), Amersham Hybond™ P (PVDF) membranes and compatible with Amersham Hyperfilm™, Amersham Hybond- ECL (nitrocellulose) membranes
  • Also compatible with Rainbow™ Molecular Weight Markers and Amersham ECL DualVue™ Western Blotting Markers

 

Education and research

Specifications

Product Type ECL Western Blot Detection Reagent
For Use With (Application) Sensitive Quantitative or Confirmatory Western Blot
Sensitivity ∼1 pg
Sufficient For 1000cm2 membrane
Content And Storage Solution A (Luminol solution, 50mL) and Solution B (Peroxide solution, 50mL), 2°C to 8°C