missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ ECL™ Blocking Agent
Description
- Contains 40g of membrane blocking agent
- Sufficient for at least 20 miniblots
Specifications
Specifications
| For Use With (Application) | Blocking Nitrocellulose and PVDF membranes to be used with ECL™ and ECL™ PlusTM western blotting detection reagents |
| Includes | 40g ECL™ blocking agent, sufficient for at least 20 miniblots |
| Quantity | 40000 mg |