missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ CyDye™ Value Packs (mono-Reactive NHS Ester)
Description
- Contains derivitized CyDyes that carry only one reactive group on each dye molecule
- pH range: 3 to 10
- DMSO tolerant
- Photostable
- Example protocols included with each dye pack
Specifications
Specifications
| For Use With (Application) | For labelling via amine groups. CyDyes are suitable for use in applications such as fluorescence resonance energy transfer (FRET), time resolved- fluorescence resonance energy transfer (TR-FRET), and fluorescence polarization (FP) |
| Quantity | 50 mg |
| Product Type | Cy5 NHS Ester |
| Kit Contents | Foil packs, each containing 1, 5, 10, 25 or 50mg dried dye |
| pH Range | 3 to 10 |
| Molecular Weight (g/mol) | 791.99 |
| Solubility | Soluble in cold water and hot water |
| Content And Storage | 2°C to 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles. |
| Label or Dye | Cy5 |
| Color | Blue |
| Show More |