missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ CyDye™ Value Packs (mono-Reactive Hydrazide)
Description
Hydrazide esters provide a convenient method for the labeling of free carbonyl groups of glycoproteins and carbohydrates. For maximum convenience, stable active hydrazide esters are supplied and stored as solid material.
- Range of bulk packs available for greatest economy
- Example protocols included with each dye pack
Specifications
Specifications
| For Use With (Application) | For labelling via carbonyl groups. For use in steady state fluorescence, FP and FRET applications |
| Quantity | 1 mg |
| Product Type | Cy3 Hydrazide |
| Kit Contents | Foil packs, each containing 1, 5 or 25mg dried dye |
| pH Range | 3 to 10 |
| Molecular Weight (g/mol) | 765.95 |
| Solubility | Soluble in cold water and hot water |
| Content And Storage | 2°C to 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles. |
| Label or Dye | Cy3 |
| Color | Red |