missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham CyDye Value Packs (bis-Reactive NHS Ester), Red
Description
Fluorescent dyes are a convenient way to label antibodies, proteins, drugs, and oligonucleotides. They are ideal for use in ELISA, immunoprecipitation and blotting applications.
- Suitable for general labeling of protein amine groups
- Used for high-throughput screening (HTS) applications
- Bulk packs for greatest economy
- Foil packs of fluors carrying two reactive groups on each dye molecule
Specifications
Specifications
| Description | Foil packs, containing dried dye. |
| For Use With (Application) | Used for high-throughput screening (HTS) applications |
| Quantity | 50 mg |
| Product Type | Cy3 Bis NHS Ester |
| Molecular Weight (g/mol) | 949.11 |
| Content And Storage | 2°C to 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles. |
| Label or Dye | Cy3 |
| Color | Red |