missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ CyDye™ Reactive Dye 5-Packs
Description
- Convenient way to label antibodies, proteins, drugs, and oligonucleotides with CyDye Fluorescent Dyes.
- For use in ELISA, immunoprecipitation, and blotting applications.
- Dyes are packaged in premixed amounts and foil-sealed to ensure consistent labelings.
Order Info
Each kit contains five vials of dye. Each vial is sufficient for labeling 1mg protein or 30nmol amino groups.
Specifications
Specifications
| For Use With (Application) | For the labelling of biological compounds with Cy5 monofunctional dye |
| Quantity | Each |
| Product Type | Cy5 Mono-Reactive Dye Pack |
| Kit Contents | Five foil packs each containing dried dye to label 1mg of Protein |
| Sufficient For | Labeling 1mg Protein or 30nM Amino groups |
| Molecular Weight (g/mol) | 791.99 |
| Content And Storage | 2°C to 8°C, protected from light |
| Label or Dye | Cy5 mono-Functional Dye |
| Color | Blue |