missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ CDP-Star Detection Reagents
Description
- Convenient for rapid separations either alone or connected in series
- Designed for use with syringe, peristaltic pump or chromatographic systems
- High flowrates and good scale-up potential
Order Info
Sufficient for detection of 2500cm2 membrane. Includes detection reagents only. For labeling module, see part RPN3680.
Specifications
Specifications
| For Use With (Application) | DNA/RNA and Probes Labeling |
| Sufficient For | 2500cm2 membrane |
| Includes | CDP-Star detection reagents |
| Quantity | Each |