missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ambroxol Hydrochloride 98.0+%, TCI America™
$143.54 - $562.95
Chemical Identifiers
| CAS | 23828-92-4 |
|---|---|
| Molecular Formula | C13H19Br2ClN2O |
| Molecular Weight (g/mol) | 414.57 |
| MDL Number | MFCD00078932 |
| InChI Key | QNVKOSLOVOTXKF-UHFFFAOYSA-N |
| Synonym | ambroxol hydrochloride, ambroxol hcl, mucoangin, mucosolvan, ambroxolhydrochloride, unii-cc995zmv90, cis-4-2-amino-3,5-dibromobenzyl amino cyclohexanol hydrochloride, trans-4-2-amino-3,5-dibromobenzyl amino cyclohexanol hydrochloride, ambroxol hydrochloride jan, lasolvan |
| PubChem CID | 108013 |
| IUPAC Name | hydrogen 4-{[(2-amino-3,5-dibromophenyl)methyl]amino}cyclohexan-1-ol chloride |
| SMILES | [H+].[Cl-].NC1=C(Br)C=C(Br)C=C1CNC1CCC(O)CC1 |
Chemical Identifiers
| 23828-92-4 | |
| 414.57 | |
| QNVKOSLOVOTXKF-UHFFFAOYSA-N | |
| 108013 | |
| [H+].[Cl-].NC1=C(Br)C=C(Br)C=C1CNC1CCC(O)CC1 |
| C13H19Br2ClN2O | |
| MFCD00078932 | |
| ambroxol hydrochloride, ambroxol hcl, mucoangin, mucosolvan, ambroxolhydrochloride, unii-cc995zmv90, cis-4-2-amino-3,5-dibromobenzyl amino cyclohexanol hydrochloride, trans-4-2-amino-3,5-dibromobenzyl amino cyclohexanol hydrochloride, ambroxol hydrochloride jan, lasolvan | |
| hydrogen 4-{[(2-amino-3,5-dibromophenyl)methyl]amino}cyclohexan-1-ol chloride |
Specifications
| 23828-92-4 | |
| White | |
| C13H19Br2ClN2O | |
| 5 g | |
| QNVKOSLOVOTXKF-UHFFFAOYSA-N | |
| hydrogen 4-{[(2-amino-3,5-dibromophenyl)methyl]amino}cyclohexan-1-ol chloride | |
| 108013 | |
| ≥98.0% (HPLC,N) | |
| Ambroxol Hydrochloride |
| 235°C | |
| 4.0 to 6.0 (1% H2O) | |
| MFCD00078932 | |
| ambroxol hydrochloride, ambroxol hcl, mucoangin, mucosolvan, ambroxolhydrochloride, unii-cc995zmv90, cis-4-2-amino-3,5-dibromobenzyl amino cyclohexanol hydrochloride, trans-4-2-amino-3,5-dibromobenzyl amino cyclohexanol hydrochloride, ambroxol hydrochloride jan, lasolvan | |
| [H+].[Cl-].NC1=C(Br)C=C(Br)C=C1CNC1CCC(O)CC1 | |
| 414.57 | |
| 414.57 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : GV8425000
TSCA : No