missing translation for 'onlineSavingsMsg'
Learn More

(-)-Ambroxide 98.0+%, TCI America™

Catalog No. A24725G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24725G 5 g
A247225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24725G Supplier TCI America Supplier No. A24725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6790-58-5
Molecular Formula C16H28O
Molecular Weight (g/mol) 236.40
MDL Number MFCD00134491
InChI Key YPZUZOLGGMJZJO-UHFFFAOYNA-N
Synonym 1,5,5,9-Tetramethyl-13-oxatricyclo[8.3.0.0(4,9)]tridecane
PubChem CID 10857465
ChEBI CHEBI:78307
IUPAC Name 3a,6,6,9a-tetramethyl-dodecahydronaphtho[2,1-b]furan
SMILES CC12CCC3C(C)(C)CCCC3(C)C1CCO2

Specifications

Melting Point 77°C
Color White
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Formula Weight 236.40
Percent Purity ≥98.0% (GC)
Chemical Name or Material (-)-Ambroxide