accessibility menu, dialog, popup

UserName

H3COH3CH3CCH3

CAS RN 6790-58-5

IUPAC Name: 3a,6,6,9a-tetramethyl-dodecahydronaphtho[2,1-b]furan
Synonyms: ambroxide
Molecular Weight (g/mol): 236.4
Molecular Formula: C16H28O
InChi Key: YPZUZOLGGMJZJO-UHFFFAOYNA-N
SMILES: CC12CCC3C(C)(C)CCCC3(C)C1CCO2
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results