missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ambrisentan 98.0+%, TCI America™
$48.13 - $136.69
Chemical Identifiers
| CAS | 177036-94-1 |
|---|---|
| Molecular Formula | C22H22N2O4 |
| Molecular Weight (g/mol) | 378.43 |
| MDL Number | MFCD08672619 |
| InChI Key | OUJTZYPIHDYQMC-UHFFFAOYNA-N |
| Synonym | (S)-2-[(4,6-Dimethylpyrimidin-2-yl)oxy]-3-methoxy-3,3-diphenylpropionic Acid |
| PubChem CID | 6918493 |
| IUPAC Name | 2-[(4,6-dimethylpyrimidin-2-yl)oxy]-3-methoxy-3,3-diphenylpropanoic acid |
| SMILES | COC(C(OC1=NC(C)=CC(C)=N1)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A296620MG
|
TCI America
A296620MG |
20 mg |
Each for $48.13
|
|
|||||
|
A2966100MG
|
TCI America
A2966100MG |
100 mg |
Each for $136.69
|
|
|||||
Chemical Identifiers
| 177036-94-1 | |
| 378.43 | |
| OUJTZYPIHDYQMC-UHFFFAOYNA-N | |
| 6918493 | |
| COC(C(OC1=NC(C)=CC(C)=N1)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1 |
| C22H22N2O4 | |
| MFCD08672619 | |
| (S)-2-[(4,6-Dimethylpyrimidin-2-yl)oxy]-3-methoxy-3,3-diphenylpropionic Acid | |
| 2-[(4,6-dimethylpyrimidin-2-yl)oxy]-3-methoxy-3,3-diphenylpropanoic acid |
Specifications
| 177036-94-1 | |
| White | |
| MFCD08672619 | |
| (S)-2-[(4,6-Dimethylpyrimidin-2-yl)oxy]-3-methoxy-3,3-diphenylpropionic Acid | |
| COC(C(OC1=NC(C)=CC(C)=N1)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 378.43 | |
| 378.43 | |
| Crystalline Powder |
| 178°C | |
| C22H22N2O4 | |
| 20 mg | |
| OUJTZYPIHDYQMC-UHFFFAOYNA-N | |
| 2-[(4,6-dimethylpyrimidin-2-yl)oxy]-3-methoxy-3,3-diphenylpropanoic acid | |
| 6918493 | |
| ≥98.0% (HPLC) | |
| Ambrisentan |
Safety and Handling
RTECSNumber : UA2459660
TSCA : No