missing translation for 'onlineSavingsMsg'
Learn More

Ambrisentan 98.0+%, TCI America™

Catalog No. A296620MG
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A296620MG 20 mg
A2966100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A296620MG Supplier TCI America Supplier No. A296620MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 177036-94-1
Molecular Formula C22H22N2O4
Molecular Weight (g/mol) 378.43
MDL Number MFCD08672619
InChI Key OUJTZYPIHDYQMC-UHFFFAOYNA-N
Synonym (S)-2-[(4,6-Dimethylpyrimidin-2-yl)oxy]-3-methoxy-3,3-diphenylpropionic Acid
PubChem CID 6918493
IUPAC Name 2-[(4,6-dimethylpyrimidin-2-yl)oxy]-3-methoxy-3,3-diphenylpropanoic acid
SMILES COC(C(OC1=NC(C)=CC(C)=N1)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 178°C
Color White
Quantity 20 mg
Formula Weight 378.43
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Ambrisentan
RTECSNumber UA2459660
TSCA No