missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMBRA1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$344.88 - $344.88
Specifications
| Antigen | AMBRA1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
Description
The recommended shelf life for this product is 1 year from date of receipt.
Ambra1, a WD40 domain containing protein, is a novel activating molecule in Beclin 1-regulated autophagy and plays a role in the development of the nervous system. The protein binds to Beclin 1 and favors Beclin 1/Vps34 interaction. The Ambra1 gene localized on human chromosome 11p11.2, encodes the protein that is highly conserved in vertebrates only and its expression is mostly confined to the neuroepithelium during early neurogenesis.Specifications
| AMBRA1 | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| DCAF3, WDR94; DDB1 and CUL4 associated factor 3; WD repeat domain 94; activating molecule in BECN1-regulated autophagy protein 1; activating molecule in beclin-1-regulated autophagy | |
| AMBRA1 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9C0C7 | |
| 55626 | |
| Between 1200 and 1250 | |
| Primary | |
| 4° C | |
| AMBRA1 |