missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Wash Solution II, RNA Purification

Catalog No. 4305890
Encompass
Change view
Click to view available options
Quantity:
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4305890 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4305890 Supplier Invitrogen™ Supplier No. 4305890
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

TransPrep Chemistry replacement solution

Applied Biosystems replacement solution for use with TransPrep Chemistry is for the isolation on the ABI PRISM™ 6100 Nucleic Acid PrepSation.

DNA and RNA Purification and Analysis, General RNA Purification Reagents and Accessories, RNA Extraction, Total RNA Isolation, Total RNA from Animal Cells and Tissues, Total RNA from Blood

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage

Store at 4°C.

1,000 mL is provided.

Buffer Wash Buffer
Quantity 1000 mL
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.